Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nick Translation System

Catalog No. 18160010
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-160-010 50 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-160-010 Supplier Invitrogen™ Supplier No. 18160010
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Nick Translation System

The Nick Translation System is ideal for both radioactive and non-radioactive labeling of DNA. The Nick Translation System:

  • Provides five pre-mixed nucleotide solutions for flexibility in labeled nucleotide incorporation
  • Yields >108 cpm/μg control DNA using [ - 32 P]-dCTP
  • Labels 1μg of DNA in one reaction
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage The Nick Translation System includes an enzyme mix containing DNA Polymerase I and DNase I, dNTPs, control DNA, stop buffer, and distilled water. Store at -20°C. Guaranteed stable for 6 months when properly stored.
    Detection Method Radioactive, Fluorescence
    Final Product Type Probes (Labeled DNA)
    Labeling Target DNA (General)
    Includes Label or Dye No
    Labeling Method Direct Labeling
    Product Type Nick Translation System
    Quantity 50 Reactions
    Shipping Condition Wet Ice
    Are labeled nucleotides included in the Nick Translation System?

    No, labeled nucleotides are not included.

    WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.