Learn More
AdipoGen Nickamine
SDP
Supplier:  AdipoGen CDXB0150M100
Encompass_Preferred
Chemical. CAS 1033772-47-2. Formula C16H20ClN3Ni. MW 348.5. Synthetic Reagent used as a catalyst for cross-coupling reactions between alkyl halides and carbon nucleophiles. Catalyst for alkyl-alkyl Kumada coupling of secondary alkyl halides, direct alkylation of heterocyclic C-H bonds, Sonogashira coupling of nonactivated alkyl halides, Kumada-Corriu-Tamao coupling of nonactivated alkyl halides with aryl and heteroaryl nucleophiles. Reactant of pincer NN2 ligand leading to selective carbon-carbon bond formation.
Catalog No. 50-256-1651