Learn More
Cpi International NICKEL 1K PPM
SDP
Catalog No. NC9947958
Encompass
Single-Element Standards 1000microgram/mL, Element: Nickel, Source: Ni metal, Purity: 99.999%, Matrix: 2% HNO3
Catalog No. NC9947958 Supplier Cpi International Supplier No. S44001000361
Add to Cart
Add to Cart
Single-Element Standards 1000microgram/mL, Element: Nickel, Source: Ni metal, Purity: 99.999%, Matrix: 2% HNO3