Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Nickel
Spécifications
Spécifications
| Melting Point | 1453°C |
| CAS | 7440-02-0 |
| Boiling Point | 2732°C |
| Density | 8.9 g/mL (at 25°C) |
| Quantity | 1 Ea. |
| Molecular Formula | Ni |
| Linear Formula | Ni |
| MDL Number | MFCD00011137 |
| Synonym | Nickel; NI000565 |
| Molecular Weight (g/mol) | 58.69 |
| Afficher plus de résultats |
missing translation for 'productTitle'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.
missing translation for 'spotOppurtunityImprovement'