Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel, AAS standard solution, Specpure™ Ni 1000μg/mL

Catalog No. AA88082AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA88082AP 500 mL
AA88082AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA88082AP Supplier Thermo Scientific Chemicals Supplier No. 088082AP
Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00011137

Specifications

Physical Form Liquid
Concentration Ni in 5% HNO3
UN Number UN3264
Color Colorless
Quantity 500 mL
Grade Specpure
Chemical Name or Material Nickel, AAS standard solution, Ni 1000μg/mL
Hazard Category H290-H314-H335-H350i-H360D
Hazard Statement GHS H Statement
H314-H317-H208
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P234-P260-P264b-P271-P281-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.