Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel Acetate Tetrahydrate (Crystalline/Certified), Fisher Chemical

Catalog No. n46500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
N46-500 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. N46-500
This item is not returnable. View return policy

Chemical Identifiers

CAS 6018-89-9
Molecular Formula C4H14NiO8
Molecular Weight (g/mol) 248.84
MDL Number MFCD00066973
InChI Key OINIXPNQKAZCRL-UHFFFAOYSA-L
Synonym nickel ii acetate tetrahydrate, nickel acetate tetrahydrate, nickel diacetate tetrahydrate, diacetatonickel tetrahydrate, nickelous acetate tetrahydrate, unii-6soa8l0560, acetic acid, nickel +2 salt, tetrahydrate, nickel 2+ diacetate tetrahydrate, nickel acetate tetrahydrate nickel and nickel compounds, acetic acid, nickel 2+ salt, tetrahydrate
PubChem CID 62601
IUPAC Name nickel(2+) diacetate tetrahydrate
SMILES O.O.O.O.[Ni++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point 250°C
Color Green
Physical Form Solid
Quantity 500 g
Linear Formula Ni(C2H3O2)2·4H2O
Formula Weight 248.84
Percent Purity 98 to 102%
Grade Certified
Identification Pass Test
Packaging Glass Bottle
Density 1.744g/cm³
Chemical Name or Material Nickel Acetate Tetrahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.