Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel Carbonate, Basic, Reagent Grade, Ricca Chemical
SDP

Catalog No. RDCN0150500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCN0150500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCN0150500 Supplier Ricca Chemical Company Supplier No. RDCN0150500A6
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 12607-70-4
Molecular Formula CH8Ni3O7
Molecular Weight (g/mol) 308.15
InChI Key JYTXCCGDBBXJAQ-UHFFFAOYSA-L
IUPAC Name trinickel(2+) tetrakis(λ3-oxidanide) carbonate
SMILES [OH2-].[OH2-].[OH2-].[OH2-].[Ni++].[Ni++].[Ni++].[O-]C([O-])=O

Specifications

Physical Form Fine Crystals or Crystalline Powder
Quantity 500 g
Grade Reagent
Identification Pass
Packaging Natural Poly Bottle
Chemical Name or Material Nickel Carbonate
ShelfLife 84 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.