Learn More
Sigma Aldrich Nickel Catalytic Static Mixer 316L 6Mm Dia X 150Mm
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
Specifications
Specifications
| Molecular Formula | C37H62N6O4 |
| Linear Formula | C37H62N6O4 |
| Quantity | 1 Ea. |
| Synonym | tert-Butyl ((S)-1-(((S)-2-((S)-2-(((S)-1-amino-13-oxo-15-phenyl-3,6,9-trioxa-12-azapentadecan-14-yl)carbamoyl)pyrrolidin-1-yl)-1-cyclohexyl-2-oxoethyl)amino)-1-oxopropan-2-yl)carbamate; AVP conjugate for IAP-mediated protein degrader development; Protein degrader building block for PROTAC(R) research |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.