Learn More
XpressBio Xpress Capture Nickel Chelate ELISA Plates, White 384 wells (5-pack)
SDP
Supplier:  XpressBio EX9000W384
Encompass_Preferred
Capture coated Nickel Chelate ELISA plates may be used with His-tagged proteins or peptides for various uses such as recombinant protein expression screening, immunoabsorbtion assays, biochemical assays, competition assays and protein purification. As little as 50 pg of His-tagged protein can be detected in a well.
Catalog No. 50-171-7661