Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel Chromium Iron powder, -325 mesh, Thermo Scientific Chemicals

Catalog No. AA44888A1
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA44888A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA44888A1 Supplier Thermo Scientific Chemicals Supplier No. 044888A1
Add to Cart

missing translation for 'validMainframeMsg'

Molecular Formula: Ni:Cr:Fe; 58:27-31:7-11 wt%

Nickel Chromium Iron powder is used for process piping, heat exchangers, carburizing equipment, heating-element sheathing, and nuclear steam-generator tubing, slitting, leveling & tension leveling, shearing & slearing, plate burning, annealing, turning & polishing, warehousing & repackaging, painting, chemical, automotive, aircraft & aerospace, medical, mechanical & food & beverage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nickel Chromium Iron powder
Quantity 1 kg
Molecular Formula Ni:Cr:Fe; 58:27-31:7-11 wt%
MDL Number MFCD01741927
UN Number UN3089
Solubility Information Insoluble in water
Packaging Glass bottle

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.