Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel Copper Iron Manganese rod, 1.27cm (0.5 in.) dia., Thermo Scientific™

Catalog No. AA36457BS
Change view
Click to view available options
Quantity:
10 cm
50 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA36457BS 10 cm
AA36457BW 50 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA36457BS Supplier Thermo Scientific Chemicals Supplier No. 36457BS
Add to Cart

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nickel Copper Iron Manganese rod
Name Note 1.27cm (0.5 in.) dia., cold drawn stress relieved, ASTM B164
Avg. Mol. Wt. or Mol. Wt. Range Ni:Cu:Fe:Mn; 63:28 to 34:2.5 max:2 max wt%
Quantity 10 cm
Weight ≈104g/10cm
Form Rod
MDL Number MFCD02091727
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.