Learn More
Strem, An Ascensus Company CAS# 7440-02-0. 100 x 30cm. Nickel foil (99%). MFCD00011137
SDP
Catalog No. 5090111205
Encompass
CAS# 7440-02-0. 100 x 30cm. Nickel foil (99%). MFCD00011137. Molecular Weight: 58.71. Molecular Formula: Ni. Color/form: 0.127mm thick x 30cm wide (~67.8g/20 x 30cm) Strem# 28-0002. http://www.strem.com/catalog/v/28-0002/
Catalog No. 50-901-11205 Supplier Strem, An Ascensus Company Supplier No. 280002100CM
CAS# 7440-02-0. 100 x 30cm. Nickel foil (99%). MFCD00011137. Molecular Weight: 58.71. Molecular Formula: Ni. Color/form: 0.127mm thick x 30cm wide (~67.8g/20 x 30cm) Strem# 28-0002. http://www.strem.com/catalog/v/28-0002/