Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel ICP Standard, 10,000 ppm Ni in 5% HNO3, Ricca Chemical
SDP

Catalog No. PNI10KN500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PNI10KN-500 500 mL Natural Poly Bottle
PNI10KN-50 50 mL Natural Poly Bottle
PNI10KN-100 100 mL Natural LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PNI10KN-500 Supplier Ricca Chemical Company Supplier No. PNI10KN500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7440-02-0 , 7697-37-2 , 7440-02-0 , 7732-18-5
Molecular Formula Ni
Molecular Weight (g/mol) 58.69
MDL Number MFCD00011137 MFCD06798735
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 10,000ppm Ni
CAS Min % 86.0, 11.57, 0.86
CAS Max % 89.0, 12.05, 0.89
Equivalence 1 mL = 10 mg Ni
Physical Form Liquid
Color Green
pH < 2
Melting Point Approximately 0°C
Type Standard
Quantity 500 mL
Chemical Name or Material Nickel ICP Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.