Learn More
SPEX CERTIPREP LLC 3P NICKEL ICP STD 100ML
Supplier:  SPEX CERTIPREP LLC 3P 1000362100
Encompass_Preferred
(HPS by Zeptometrix) 1000 ug/mL Nickel in 2% HNO3, 100mL, sourced from Ni metal, 18 mo expiry date, This standard is traceable to NIST and certified Guide 34 and 17025.
Catalog No. NC0216591
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.