Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel(II) 2,4-pentanedionate hydrate, Thermo Scientific™

Catalog No. AA12529A1
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA12529A1 1 kg
AA1252930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA12529A1 Supplier Thermo Scientific Chemicals Supplier No. 12529A1

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120156-44-7
Molecular Formula C10H14NiO4
Molecular Weight (g/mol) 256.91
MDL Number MFCD00000024
InChI Key BMGNSKKZFQMGDH-FDGPNNRMSA-L
Synonym 3z-4-2z-4-oxopent-2-en-2-yl oxy nickelio oxy pent-3-en-2-one hydrate, c10h14nio4.h2o, nickel ii acetylacetonate hydrate, acetylacetone nickel ii salt hydrate, z-4-hydroxypent-3-en-2-one;nickel;hydrate
PubChem CID 91886337
IUPAC Name nickel(2+) bis((2Z)-4-oxopent-2-en-2-olate)
SMILES [Ni++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Boiling Point 220°C (11mmHg)
Melting Point ∼230°C (decomposition)
Physical Form Powder
Quantity 1 kg
Beilstein 4157970
Merck Index 14,6500
Formula Weight 256.91 (Anhydrous)
Chemical Name or Material Nickel(II) 2,4-pentanedionate hydrate
Hazard Category Warning
Hazard Statement GHS H Statement
H351
Suspected of causing cancer.
Precautionary Statement GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECSNumber SA2100000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.