Learn More
Strem, An Ascensus Company CAS# 28923-39-9. Nickel(II) bromide, dimethoxyethane adduct, min. 97%. MFCD00013480. 1G.
SDP
Supplier:  Strem, An Ascensus Company 2811451G
Encompass
CAS# 28923-39-9. 1G. Nickel(II) bromide, dimethoxyethane adduct, min. 97%. MFCD00013480. Molecular Formula: C4H10Br2NiO2. Molecular Weight: 308.62. Color/Form: peach pwdr. Stability: moisture sensitive Strem#: 28-1145 www.strem.com/catalog/v3/28-1145/
Catalog No. 50-195-9528