Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Nickel(II) 5,9,14,18,23,27,32,36-octabutoxy-2,3-naphthalocyanine

Catalog No. 418854100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
Catalog No. Quantity
41-885-4100MG 100 mg
1 options

Catalog No. 41-885-4100MG

Supplier: Sigma Aldrich 418854100MG

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Nickel(II) 5,9,14,18,23,27,32,36-octabutoxy-2,3-naphthalocyanine

Specifications

CAS 155773-70-9
Melting Point >300°C
Molecular Formula C80H88N8NiO8
Linear Formula C80H88N8NiO8
MDL Number MFCD00192345
Quantity 100 mg
Synonym 5,9,14,18,23,27,32,36-Octabutoxy-2,3-naphthalocyanine nickel(II) salt
Molecular Weight (g/mol) 1348.3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.