Learn More
Strem, An Ascensus Company CAS# 6018-89-9. 250g. Nickel(II) acetate tetrahydrate, 98+%. MFCD00066973
SDP
Catalog No. 5090114771
Encompass
CAS# 6018-89-9. 250g. Nickel(II) acetate tetrahydrate, 98+%. MFCD00066973. Molecular Weight: 176.80 (248.86). Molecular Formula: Ni(OOCCH3)2∙4H2O. Color/form: green xtl. Strem# 93-2803. http://www.strem.com/catalog/v/93-2803/
Catalog No. 50-901-14771 Supplier Strem, An Ascensus Company Supplier No. 932803250G
CAS# 6018-89-9. 250g. Nickel(II) acetate tetrahydrate, 98+%. MFCD00066973. Molecular Weight: 176.80 (248.86). Molecular Formula: Ni(OOCCH3)2∙4H2O. Color/form: green xtl. Strem# 93-2803. http://www.strem.com/catalog/v/93-2803/