Alfa Aesar™ Nickel(II) acetate tetrahydrate, Puratronic™, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 1081306

Quantity 5g

 View more versions of this product

Catalog No. AA1081306

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 6018-89-9
    Linear Formula Ni(CH3COO)2·4H2O
    Formula Weight 248.84 (176.79 Anhydrous)
    MDL Number MFCD00066973
    UN Number UN3077
    Beilstein 3730764
    Merck Index 14,6499
    Chemical Name or Material Nickel(II) acetate tetrahydrate
    Grade Puratronic™

    Specifications

    Formula Weight 248.84 (176.79 Anhydrous)
    Health Hazard 2 GHS H Statement
    H334-H350-H360-H372-H341-H302-H332-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause cancer.
    May damage fertility or the unborn child.
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing genetic defects.
    Harmful if swallowed.
    Harmful if inhaled.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P260-P284a-P261-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Beilstein 3730764
    Odor Light
    MDL Number MFCD00066973
    Color Green
    Health Hazard 1 Danger
    Density 1.744g/mL
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    TSCA TSCA
    Linear Formula Ni(CH3COO)2·4H2O
    CAS 6018-89-9
    UN Number UN3077
    Merck Index 14,6499
    Quantity 5g
    Physical Form Crystalline Aggregates
    RTECS Number QR6126000
    Chemical Name or Material Nickel(II) acetate tetrahydrate
    Assay 99.999%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H334-H350-H360-H372-H341-H302-H332-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause cancer.
    May damage fertility or the unborn child.
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing genetic defects.
    Harmful if swallowed.
    Harmful if inhaled.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P284a-P261-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit