Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Nickel(II) bromide ethylene glycol dimethyl ether complex

Catalog No. 4063415G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-634-15G 5 g
40-634-11G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 40-634-15G Supplier Sigma Aldrich Supplier No. 4063415G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Nickel(II) bromide ethylene glycol dimethyl ether complex is a nickel based catalyst that is used in the preparation of poly(3-hexylthiophene) (P3HT) Kumada catalyst transfer polymerization. It facilitates good control over the molecular weight of the conjugating polymer.

Specifications

CAS 28923-39-9
Molecular Formula C4H10Br2NiO2
Linear Formula NiBr2 -+ CH3OCH2CH2OCH3
MDL Number MFCD00013480
Quantity 5 g
Synonym (1,2-Dimethoxyethane)nickel dibromide; Dibromo(1,2-dimethoxyethane)nickel; Dibromo(1,2-dimethoxyethane)nickel(II)
Molecular Weight (g/mol) 308.62
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.