Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Nickel(II) chloride

Catalog No. 33935050G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-935-050G 50 g
33-935-0250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 33-935-050G Supplier Sigma Aldrich Supplier No. 33935050G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Nickel(II) chloride (NiCl2) is a nickel based halide that is prepared by burning nickel in chlorine. It is a water soluble compound that crystallizes to form a hexahydrate. It is majorly utilized in organic synthesis as a catalyst and a precursor.NiCl2 can be used in the form of a nanoparticle for the development of separation membranes for the wastewater treatment. It can also be used to form an electron with sodium ions for the fabrication of sodium nickel chloride based batteries.

Specifications

CAS 7718-54-9
Density 3.55 g/mL (at 25°C)
Molecular Formula Cl2Ni
Linear Formula NiCl2
Quantity 50 g
MDL Number MFCD00011142
Synonym Nickel dichloride; Nickel(2+) chloride; Nickelous chloride
Molecular Weight (g/mol) 129.6
Percent Purity 98%
Biological Activity Lungs
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.