Learn More
Strem, An Ascensus Company CAS# 7791-20-0. 250g. Nickel(II) chloride hexahydrate (99.9+%-Ni). MFCD00149809
SDP
Catalog No. 5090114780
Encompass
CAS# 7791-20-0. 250g. Nickel(II) chloride hexahydrate (99.9+%-Ni). MFCD00149809. Molecular Weight: 129.62 (237.70). Molecular Formula: NiCl2∙6H2O. Color/form: green xtl. Strem# 93-2810. http://www.strem.com/catalog/v/93-2810/
Catalog No. 50-901-14780 Supplier Strem, An Ascensus Company Supplier No. 932810250G
CAS# 7791-20-0. 250g. Nickel(II) chloride hexahydrate (99.9+%-Ni). MFCD00149809. Molecular Weight: 129.62 (237.70). Molecular Formula: NiCl2∙6H2O. Color/form: green xtl. Strem# 93-2810. http://www.strem.com/catalog/v/93-2810/