Learn More
Strem, An Ascensus Company CAS# 7791-20-0. 10g. Nickel(II) chloride hexahydrate (99.999+%-Ni) PURATREM. MFCD00149809
SDP
Supplier:  Strem, An Ascensus Company 28116010G
Encompass
CAS# 7791-20-0. 10g. Nickel(II) chloride hexahydrate (99.999+%-Ni) PURATREM. MFCD00149809. Molecular Weight: 129.62 (237.70). Molecular Formula: NiCl2∙6H2O. Color/form: green xtl. Strem# 28-1160. http://www.strem.com/catalog/v/28-1160/
Catalog No. 50-901-11253