Learn More
Nickel(II) chloride hydrate, Puratronic™, 99.995% (metals basis)
Description
It is used in electroplating and as a catalyst for organic conversions, for example in chemo-selective thioacetalization of aldehydes. In combination with lithium aluminum hydride, it serves as a reducing agent for alkenes, alkynes, and organic halides; it can cleave N-O bond and open epoxides. It is a precursor to several nickel-phosphine complexes, such as bis(triphenylphosphine)nickel(II) chloride, which are used in alkyne trimerizations, carbonylations, and as catalysts in organic reactions such as Suzuki-Miyaura cross coupling reactions as an alternative to palladium(0) catalysts. It is the precursor to acetylacetonate complex of Ni, used for producing 1,5-cyclooctadiene complex, an important reagent in organonickel chemistry. It can be used to prepare the sandwich compound nickelocene through dimethoxyethane complex of nickel chloride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Green |
| Physical Form | Crystalline Aggregates |
| Quantity | 5 g |
| Linear Formula | NiCl2·xH2O |
| UN Number | UN3288 |
| Merck Index | 14,6505 |
| Solubility Information | Freely soluble in water. Soluble in ethylene glycol,ethanol,and ammonium hydroxide. |
| IUPAC Name | dichloronickel;hydrate |
| Formula Weight | 129.60 (Anhydrous) |
| Percent Purity | 99.995% |
| Show More |
Safety and Handling
| Hazard Category | H301+H331-H315-H317-H319-H334-H341-H350i-H360D-H372 |
| Hazard Statement | GHS H Statement H301-H331-H334-H350-H360-H372-H341-H315-H317 Toxic if swallowed. Toxic if inhaled. May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause cancer. May damage fertility or the unborn child. Causes damage to organs through prolonged or repeated exposure. Suspected of causing genetic defects. Causes skin irritation. May cause an allergic skin reaction. |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P330-P333+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S. |
| EINECSNumber | 231-743-0 |
| RTECSNumber | QR6480000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.