Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel(II) chloride, ultra dry, 99.9% (metals basis)

Catalog No. AA3571306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3571306 5 g
AA3571314 25 g
AA3571322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3571306 Supplier Thermo Scientific Chemicals Supplier No. 03571306
Add to Cart

CAS: 7718-54-9 | Cl2Ni | 129.593 g/mol

It is used in the synthesis of semiconducting metal-containing polymers in which the polypyrrole backbone has a conformational energy minimum and is nearly planar.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7718-54-9
Molecular Formula Cl2Ni
Molecular Weight (g/mol) 129.593
MDL Number MFCD00011142
InChI Key QMMRZOWCJAIUJA-UHFFFAOYSA-L
PubChem CID 24385
ChEBI CHEBI:34887
IUPAC Name dichloronickel
SMILES Cl[Ni]Cl

Specifications

Melting Point 1001°C
Color Yellow to Orange
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.9% (metals basis)
Solubility Information Soluble in water.
Formula Weight 129.6
Odor Odorless
Sensitivity Hygroscopic
Appearance Yellow to orange powder
Vapor Pressure 1.33 hPa (1mm Hg) at 20°C
Density 3.55
Chemical Name or Material Nickel(II) chloride
Show More Show Less
Hazard Category H301+H331-H315-H317-H319-H334-H341-H350i-H360D-H372-H500
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P330-P333+P313-P362-P501c
DOTInformation DOT Class: 6.1; Packing Group: III
EINECSNumber 231-743-0
RTECSNumber QR6475000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.