Learn More
Nickel(II) chloride, ultra dry, 99.9% (metals basis)
Description
It is used in the synthesis of semiconducting metal-containing polymers in which the polypyrrole backbone has a conformational energy minimum and is nearly planar.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1001°C |
| Color | Yellow to Orange |
| Physical Form | Powder |
| Quantity | 100 g |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Appearance | Yellow to orange powder |
| Vapor Pressure | 1.33 hPa (1mm Hg) at 20°C |
| Assay Percent Range | 99.9% (metals basis) |
| Density | 3.55 |
| Show More |
Safety and Handling
| Hazard Category | H301+H331-H315-H317-H319-H334-H341-H350i-H360D-H372-H500 |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P330-P333+P313-P362-P501c |
| DOTInformation | DOT Class: 6.1; Packing Group: III |
| EINECSNumber | 231-743-0 |
| RTECSNumber | QR6475000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.