Learn More
Medchemexpress LLC Nickel(II) dibutyldithiocarbamate | 13927-77-0 | MFCD00067273 | 95.0% | 467.45 | C18H36N2NiS4 | 500 G
SDP
Catalog No. 5000299351
Encompass_Preferred
Nickel(II) dibutyldithiocarbamate is an organometallic antioxidant and antiozonant used in research and industrial chemistry. It is employed in studies of oxidative stress and to protect plant tissues from ozone-induced bronzing, and it serves as a reagent in metal-dithiocarbamate chemistry.
  • Acts as an antioxidant and antiozonant
  • Useful reagent for ozone-protection and oxidative stress studies
  • High purity: 95.0%
  • Chemical formula C18H36N2NiS4 and molecular weight 467.45 aid identification
  • Available in multiple pack sizes for bench or bulk use
Catalog No. 50-002-99351 Supplier Medchemexpress LLC Supplier No. HYW035106500G
Nickel(II) dibutyldithiocarbamate is an organometallic antioxidant and antiozonant used in research and industrial chemistry. It is employed in studies of oxidative stress and to protect plant tissues from ozone-induced bronzing, and it serves as a reagent in metal-dithiocarbamate chemistry.
  • Acts as an antioxidant and antiozonant
  • Useful reagent for ozone-protection and oxidative stress studies
  • High purity: 95.0%
  • Chemical formula C18H36N2NiS4 and molecular weight 467.45 aid identification
  • Available in multiple pack sizes for bench or bulk use