Learn More
Medchemexpress LLC Nickel(II) dibutyldithiocarbamate | 13927-77-0 | MFCD00067273 | 95.0% | 467.45 g/mol | C18H36N2NiS4 | 100 G
SDP
Catalog No. 5000300240
Encompass_Preferred
Nickel(II) dibutyldithiocarbamate is an organometallic antioxidant and antiozonant supplied as a light green solid for research use. It is used in studies of oxidative processes, metal-dithiocarbamate complexes, and ozone-induced bronzing in plant tissues. The supplier provides purity and storage recommendations for reliable handling.
  • Antioxidant and antiozonant activity for oxidative-stress research.
  • Research-grade purity suitable for laboratory assays.
  • Light green solid form for convenient storage and weighing.
  • Stable when stored protected from light and under inert atmosphere.
  • Suitable for studies of metal-ligand interactions and plant physiology.
Catalog No. 50-003-00240 Supplier Medchemexpress LLC Supplier No. HYW035106100G
Nickel(II) dibutyldithiocarbamate is an organometallic antioxidant and antiozonant supplied as a light green solid for research use. It is used in studies of oxidative processes, metal-dithiocarbamate complexes, and ozone-induced bronzing in plant tissues. The supplier provides purity and storage recommendations for reliable handling.
  • Antioxidant and antiozonant activity for oxidative-stress research.
  • Research-grade purity suitable for laboratory assays.
  • Light green solid form for convenient storage and weighing.
  • Stable when stored protected from light and under inert atmosphere.
  • Suitable for studies of metal-ligand interactions and plant physiology.