Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nickel(II) dibutyldithiocarbamate | 13927-77-0 | MFCD00067273 | 95.0% | 467.45 g·mol⁻¹ | C18H36N2NiS4 | 50 G
SDP

Catalog No. 5000299305
Encompass_Preferred

Nickel(II) dibutyldithiocarbamate is a nickel coordination compound used as an antioxidant and antiozonant in research and agricultural studies. It is reported to protect plant tissues from ozone-induced bronzing and is supplied as a solid research reagent suitable for laboratory use.

  • High purity (95.0%).
  • Molecular formula C18H36N2NiS4; molecular weight 467.45 g·mol⁻¹.
  • Appearance: light green solid, convenient handling for solid-phase work.
  • Storage: protect from light; refrigerate solid; solutions may be stored at -80°C (long term) or -20°C (short term).
  • Applications: antioxidant and antiozonant studies, plant physiology experiments, and coordination chemistry research.

Catalog No. 50-002-99305 Supplier Medchemexpress LLC Supplier No. HYW03510650G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nickel(II) dibutyldithiocarbamate is a nickel coordination compound used as an antioxidant and antiozonant in research and agricultural studies. It is reported to protect plant tissues from ozone-induced bronzing and is supplied as a solid research reagent suitable for laboratory use.

  • High purity (95.0%).
  • Molecular formula C18H36N2NiS4; molecular weight 467.45 g·mol⁻¹.
  • Appearance: light green solid, convenient handling for solid-phase work.
  • Storage: protect from light; refrigerate solid; solutions may be stored at -80°C (long term) or -20°C (short term).
  • Applications: antioxidant and antiozonant studies, plant physiology experiments, and coordination chemistry research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.