Learn More
Medchemexpress LLC Nickel(II) dibutyldithiocarbamate | 13927-77-0 | MFCD00067273 | >97.0% | 467.5 g/mol | C18H36N2NiS4 | 25 G
SDP
Catalog No. 5000303042
Encompass_Preferred
Nickel(II) dibutyldithiocarbamate is an organometallic complex used as an antioxidant and antiozonant and as an accelerator and stabilizer in polymer and rubber applications. It is supplied as a research and reagent chemical with molecular formula C18H36N2NiS4, molecular weight 467.5 g/mol, and CAS 13927-77-0.
  • Acts as an antioxidant and antiozonant for synthetic rubbers.
  • Functions as a vulcanization accelerator and stabilizer.
  • Serves as a polymerization promoter and intermediate in chemical synthesis.
  • Supplied as a reagent for laboratory and industrial research use.
  • High purity (>97.0%) supports reproducible results.
  • Well-documented physical and chemical identifiers for regulatory use.
Catalog No. 50-003-03042 Supplier Medchemexpress LLC Supplier No. 25GHYW035106
Nickel(II) dibutyldithiocarbamate is an organometallic complex used as an antioxidant and antiozonant and as an accelerator and stabilizer in polymer and rubber applications. It is supplied as a research and reagent chemical with molecular formula C18H36N2NiS4, molecular weight 467.5 g/mol, and CAS 13927-77-0.
  • Acts as an antioxidant and antiozonant for synthetic rubbers.
  • Functions as a vulcanization accelerator and stabilizer.
  • Serves as a polymerization promoter and intermediate in chemical synthesis.
  • Supplied as a reagent for laboratory and industrial research use.
  • High purity (>97.0%) supports reproducible results.
  • Well-documented physical and chemical identifiers for regulatory use.