Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nickel(II) fluoride | 10028-18-9 | 99.0% | 96.69 g/mol | NiF2 | 5 G
SDP

Catalog No. 5000208205
Encompass_Preferred

Nickel(II) fluoride is an inorganic nickel fluoride used as a fluorinating agent and catalyst in surface treatments and specialty chemical syntheses. It is supplied as a high-purity solid suitable for research and industrial applications.

  • Chemical formula: NiF2.
  • CAS number: 10028-18-9.
  • Molecular weight: 96.69 g/mol.
  • Purity: 99.0%.
  • Typical packaging: 5 g, 10 g, 25 g, 50 g, 100 g.
  • Common uses: fluorinating agent, catalyst, surface treatment.

Catalog No. 50-002-08205 Supplier Medchemexpress LLC Supplier No. HYW0757665G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nickel(II) fluoride is an inorganic nickel fluoride used as a fluorinating agent and catalyst in surface treatments and specialty chemical syntheses. It is supplied as a high-purity solid suitable for research and industrial applications.

  • Chemical formula: NiF2.
  • CAS number: 10028-18-9.
  • Molecular weight: 96.69 g/mol.
  • Purity: 99.0%.
  • Typical packaging: 5 g, 10 g, 25 g, 50 g, 100 g.
  • Common uses: fluorinating agent, catalyst, surface treatment.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.