Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nickel(II) fluoride | 10028-18-9 | MFCD00016263 | 99.0% | 96.69 g/mol | NiF2 | 25 G
SDP

Catalog No. 5000208897
Encompass_Preferred

Nickel(II) fluoride is an inorganic fluorinating agent and catalyst used in surface treatment, inks, fluorescent lamp production, and organic synthesis. It is supplied as a reagent for research use.

  • Inorganic fluorinating agent and catalyst used in surface treatment and organic synthesis.
  • High purity suitable for research applications.
  • Solubility in water about 3.33 mg/mL with ultrasonic heating to 60°C.
  • Typical storage: 4°C, sealed and protected from moisture.
  • Molecular weight 96.69 g/mol; CAS 10028-18-9.

Catalog No. 50-002-08897 Supplier Medchemexpress LLC Supplier No. 25GHYW075766
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nickel(II) fluoride is an inorganic fluorinating agent and catalyst used in surface treatment, inks, fluorescent lamp production, and organic synthesis. It is supplied as a reagent for research use.

  • Inorganic fluorinating agent and catalyst used in surface treatment and organic synthesis.
  • High purity suitable for research applications.
  • Solubility in water about 3.33 mg/mL with ultrasonic heating to 60°C.
  • Typical storage: 4°C, sealed and protected from moisture.
  • Molecular weight 96.69 g/mol; CAS 10028-18-9.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.