Learn More
Nickel(II) formate, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Quantity | 100 g |
| Linear Formula | Ni(HCO2)2 |
| UN Number | UN2811 |
| Beilstein | 14,6509 |
| Synonym | 4-methylumbelliferyl phosphate, 4-methyl-2-oxo-2h-chromen-7-yl dihydrogen phosphate, 4-mup, 2h-1-benzopyran-2-one, 4-methyl-7-phosphonooxy, phosphoric acid 4-methylumbelliferyl ester, 4-methylumbelliferylphosphate, free acid, 4-methyl-2-oxochromen-7-yl oxyphosphonic acid, 5,5-dinitrobarbutyric acid, 4-methylumbelliferone phosphate, 4-methylumbelliferyl phosphate, free acid |
| SMILES | [Ni++].[O-]C=O.[O-]C=O |
| IUPAC Name | nickel(2+) diformate |
| PubChem CID | 65118 |
| Formula Weight | 148.74 |
| Chemical Name or Material | Nickel (II) formate |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement H334-H350-H360-H372-H341-H317 May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause cancer. May damage fertility or the unborn child. Causes damage to organs through prolonged or repeated exposure. Suspected of causing genetic defects. May cause an allergic skin reaction. |
| Precautionary Statement | GHS P Statement P260-P284a-P261-P342+P311-P405-P501a Do not breathe dust/fume/gas/mist/vapors/spray. Avoid breathing dust/fume/gas/mist/vapors/spray. If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
| TSCA | TSCA |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.