Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel(II) formate, Thermo Scientific™

Catalog No. AAA1155736
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1155736 500 g
AAA1155722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1155736 Supplier Thermo Scientific Chemicals Supplier No. A1155736

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3349-06-2
Molecular Formula C2H2NiO4
Molecular Weight (g/mol) 148.73
MDL Number MFCD00050833
InChI Key HZPNKQREYVVATQ-UHFFFAOYSA-L

Specifications

Quantity 500 g
Linear Formula Ni(HCO2)2
UN Number UN2811
Beilstein 14,6509
Synonym nickel formate, nickel diformate, nickel ii formate, unii-y4yt27n8qn, nickel 2+ diformate, y4yt27n8qn, formic acid, nickel 2+ salt, formic acid, nickel salt, formic acid, nickel 2+ salt 2:1, 64-18-6 parent
SMILES C(=O)[O-].C(=O)[O-].[Ni+2]
IUPAC Name nickel(2+);diformate
PubChem CID 27506
Formula Weight 148.74
Chemical Name or Material Nickel (II) formate
Hazard Category Danger
Hazard Statement GHS H Statement
H334-H350-H360-H372-H341-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
P260-P284a-P261-P342+P311-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.