Learn More
Strem, An Ascensus Company CAS# 12054-48-7. 250g. Nickel(II) hydroxide. MFCD00011140
SDP
Catalog No. 5090114823
Encompass
CAS# 12054-48-7. 250g. Nickel(II) hydroxide. MFCD00011140. Molecular Weight: 92.72. Molecular Formula: Ni(OH)2. Color/form: green pwdr. Strem# 93-2847. http://www.strem.com/catalog/v/93-2847/
Catalog No. 50-901-14823 Supplier Strem, An Ascensus Company Supplier No. 932847250G
CAS# 12054-48-7. 250g. Nickel(II) hydroxide. MFCD00011140. Molecular Weight: 92.72. Molecular Formula: Ni(OH)2. Color/form: green pwdr. Strem# 93-2847. http://www.strem.com/catalog/v/93-2847/