Learn More
Strem, An Ascensus Company CAS# 13462-90-3. 10g. Nickel(II) iodide, anhydrous (99.5%-Ni). MFCD00016264
SDP
Catalog No. 5090114797
Encompass
CAS# 13462-90-3. 10g. Nickel(II) iodide, anhydrous (99.5%-Ni). MFCD00016264. Molecular Weight: 312.52. Molecular Formula: NiI2. Color/form: -60 mesh black pwdr. Strem# 93-2827. http://www.strem.com/catalog/v/93-2827/
Catalog No. 50-901-14797 Supplier Strem, An Ascensus Company Supplier No. 93282710G
CAS# 13462-90-3. 10g. Nickel(II) iodide, anhydrous (99.5%-Ni). MFCD00016264. Molecular Weight: 312.52. Molecular Formula: NiI2. Color/form: -60 mesh black pwdr. Strem# 93-2827. http://www.strem.com/catalog/v/93-2827/