Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel(II) Nitrate Hexahydrate, purum p.a, ≥97.0% (KT), Solstice

Catalog No. 60047425 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-425 1 kg Poly bottle
60-047-426 250 g Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-047-425 Supplier Solstice Supplier No. 722531KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Purum p.a., crystallized,≥97.0%

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13478-00-7 , 7732-18-5
Molecular Formula H12N2NiO12
Molecular Weight (g/mol) 290.79
MDL Number MFCD00149805
InChI Key AOPCKOPZYFFEDA-UHFFFAOYSA-N
Synonym nickel nitrate hexahydrate, nickel ii nitrate hexahydrate, nickel dinitrate hexahydrate, nickel 2+ dinitrate hexahydrate, unii-xbt61wlt1j, nickelous nitrate hexahydrate, nickel 2+ nitrate, hexahydrate, xbt61wlt1j, nitric acid, nickel 2+ salt, hexahydrate, nickel ii nitrate, hexahydrate 1:2:6
PubChem CID 61630
SMILES O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 56.7°C
Color Blue Green
pH 4.0 (at 20°C)
Physical Form Solid
Quantity 1 kg
Packaging Poly bottle
Density 2.05g/mL at 25°C
Linear Formula Ni(NO3)2 · 6H2O
UN Number UN2725
Solubility Information 2.385g/L at 20°C
IUPAC Name nickel(2+);dinitrate;hexahydrate
Formula Weight 290.79g/mol
Percent Purity ≥97.0% (KT)
Grade purum p.a.
Chemical Name or Material Nickel(II) nitrate hexahydrate
Show More Show Less
Hazard Statement P201-P210-P261-P273-P280-P284
DOTInformation 5.1 / PGIII
RTECSNumber 236-068-5
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.