Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel(II) oxide, black, 76% Ni

Catalog No. AC223121000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC223121000 100 g Glass bottle
AC223125000 500 g Glass bottle
AC223120025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC223121000 Supplier Thermo Scientific Chemicals Supplier No. 223121000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 1313-99-1 | NiO | 74.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-99-1
Molecular Formula NiO
Molecular Weight (g/mol) 74.69
MDL Number MFCD00011145
InChI Key GNRSAWUEBMWBQH-UHFFFAOYSA-N
Synonym nickel ii oxide, nickel monoxide, bunsenite, nickelous oxide, nickel oxide, mononickel oxide, nickel protoxide, black nickel oxide, nickel 2+ oxide
PubChem CID 14805
IUPAC Name oxonickel
SMILES O=[Ni]

Specifications

Color Black-Gray
Physical Form Powder
Quantity 100 g
Linear Formula NiO
Merck Index 15, 6597
Formula Weight 74.7
Percent Purity 75 to 77% (Ni)
Packaging Glass bottle
Density 6.67g/mL
Chemical Name or Material Nickel(II) oxide, black, 76% Ni
missing translation for 'healthHazard2' GHS H Statement:
May cause cancer by inhalation.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
May cause long lasting harmful effects to aquatic life.
missing translation for 'healthHazard3' GHS P Statement:
Wear eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 215-215-7
RTECSNumber QR8400000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.