Learn More
Strem, An Ascensus Company CAS# 1313-99-1. 250g. Nickel(II) oxide, green, 99%. MFCD00011145
SDP
Catalog No. 5090114808
Encompass
CAS# 1313-99-1. 250g. Nickel(II) oxide, green, 99%. MFCD00011145. Molecular Weight: 74.71. Molecular Formula: NiO. Color/form: -325 mesh green pwdr. Strem# 93-2832. http://www.strem.com/catalog/v/93-2832/
Catalog No. 50-901-14808 Supplier Strem, An Ascensus Company Supplier No. 932832250G
CAS# 1313-99-1. 250g. Nickel(II) oxide, green, 99%. MFCD00011145. Molecular Weight: 74.71. Molecular Formula: NiO. Color/form: -325 mesh green pwdr. Strem# 93-2832. http://www.strem.com/catalog/v/93-2832/