Learn More
Nickel(II) perchlorate hexahydrate, Reagent Grade
Description
Nickel(II) perchlorate hexahydrate is used as a starting material for homometallic, trinuclear heteroscorpionate complexes applied in studies of electronic and magnetic properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 140°C |
| Color | Green |
| Physical Form | Damp Crystalline Solid |
| Quantity | 100 g |
| Solubility Information | Very soluble in water. Soluble in many organic solvents |
| Formula Weight | 365.70 (257.59 Anhydrous) |
| Grade | Reagent |
| Odor | Odorless |
| Refractive Index | 1.55 |
| Sensitivity | Hygroscopic |
| Show More |
Safety and Handling
| Hazard Category | H272-H314-H317-H334-H335-H341-H350-H360-H372 |
| Hazard Statement | Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets. |
| Precautionary Statement | P201-P202-P210-P220-P221-P260-P264b-P270-P271-P272-P280g-P281-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P370+P378q-P501c |
| DOTInformation | DOT Class: 5.1, Packaging Group: II |
| EINECSNumber | 237-124-1 |
| RTECSNumber | SC9550000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.