Learn More
Strem, An Ascensus Company CAS# 10101-97-0. 100g. Nickel(II) sulfate hexahydrate (99.99%-Ni) PURATREM. MFCD00149813
SDP
Catalog No. 501385559
Encompass
CAS# 10101-97-0. 100g. Nickel(II) sulfate hexahydrate (99.99%-Ni) PURATREM. MFCD00149813. Molecular Weight: 154.78 (262.86). Molecular Formula: NiSO4∙6H2O. Color/form: green xtl. Strem# 93-2856. http://www.strem.com/catalog/v/93-2856/
Catalog No. 50-138-5559 Supplier Strem, An Ascensus Company Supplier No. 932856100G
CAS# 10101-97-0. 100g. Nickel(II) sulfate hexahydrate (99.99%-Ni) PURATREM. MFCD00149813. Molecular Weight: 154.78 (262.86). Molecular Formula: NiSO4∙6H2O. Color/form: green xtl. Strem# 93-2856. http://www.strem.com/catalog/v/93-2856/