Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Nickel(II) sulfate hexa-/ heptahydrate

Catalog No. 1363525KGR Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-635-25KGR 2.5 kg
13-635-1KGR 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-635-25KGR Supplier Sigma Aldrich Supplier No. 136352.5KGR
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Nickel(II) sulfate hexa-/ heptahydrate

Specifications

Quantity 2.5 kg
Molecular Formula NiO4S -+ xH2O
Linear Formula NiSO4 -+ 6-7H2O
MDL Number MFCD00011146
Molecular Weight (g/mol) 154.76
Percent Purity ≥20.6%Ni and Co basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.