Learn More
Strem, An Ascensus Company CAS# 10101-97-0. 500g. Nickel(II) sulfate hexahydrate, 98+% (ACS). MFCD00149813
SDP
Catalog No. 5090114813
Encompass
CAS# 10101-97-0. 500g. Nickel(II) sulfate hexahydrate, 98+% (ACS). MFCD00149813. Molecular Weight: 154.78 (262.86). Molecular Formula: NiSO4∙6H2O. Color/form: green xtl. Strem# 93-2839. http://www.strem.com/catalog/v/93-2839/
Catalog No. 50-901-14813 Supplier Strem, An Ascensus Company Supplier No. 932839500G
CAS# 10101-97-0. 500g. Nickel(II) sulfate hexahydrate, 98+% (ACS). MFCD00149813. Molecular Weight: 154.78 (262.86). Molecular Formula: NiSO4∙6H2O. Color/form: green xtl. Strem# 93-2839. http://www.strem.com/catalog/v/93-2839/