Learn More
Sigma Aldrich Fine Chemicals Biosciences Nickel(II) sulfate hexahydrate ReagentPlus(R), powder or crystals | 10101-97-0 | MFCD00149813 | 1KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences N48821KG
Encompass
Nickel(II) sulfate hexahydrate ReagentPlus(R), powder or crystals | Purity: 98.0-102.0% (EDTA titration) | Mol Wt: 262.85 | 10101-97-0 | MFCD00149813 | 1KG
Catalog No. 50-187-9222