Learn More
Strem, An Ascensus Company CAS# 15684-36-3. 50g. Nickel(II) tetrafluoroborate hexahydrate, 99%. MFCD00150259
SDP
Catalog No. NC1512279
Encompass
CAS# 15684-36-3. 50g. Nickel(II) tetrafluoroborate hexahydrate, 99%. MFCD00150259. Molecular Weight: 232.23 (340.32). Molecular Formula: Ni(BF4)2∙6H2O. Color/form: green xtl. Strem# 93-2841. http://www.strem.com/catalog/v/93-2841/
Catalog No. NC1512279 Supplier Strem, An Ascensus Company Supplier No. 93284150G
CAS# 15684-36-3. 50g. Nickel(II) tetrafluoroborate hexahydrate, 99%. MFCD00150259. Molecular Weight: 232.23 (340.32). Molecular Formula: Ni(BF4)2∙6H2O. Color/form: green xtl. Strem# 93-2841. http://www.strem.com/catalog/v/93-2841/