Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel(II) tetrafluoroborate hexahydrate

Catalog No. p-7080289
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA5310822 100 g
AA5310836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA5310822 Supplier Thermo Scientific Chemicals Supplier No. 05310822
Only null left
Add to Cart

CAS: 15684-36-3 | B2F8H12NiO6 | 340.39 g/mol

It is mainly used as an electroplating intermediate. It is also used in medical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15684-36-3
Molecular Formula B2F8H12NiO6
Molecular Weight (g/mol) 340.39
MDL Number MFCD00150259
InChI Key BFFWGFMAXHCLDV-UHFFFAOYSA-N
Synonym nickel ii tetrafluoroborate hexahydrate, nickel ii ditetrafluoroborate hexahydrate, nickel 2+ ditetrafluoroborate hexahydrate, borate 1-,tetrafluoro-, nickel 2+ 2:1 , hexahydrate 9ci, 2bf4.ni.6h2o, nickel ii tetrafluoroboratehexahydrate, nickel ii tetrafluoroborate hexahydrate 100g, nickel 2+ hexahydrate ditetrafluoroborate, nickel 2+ ion hexahydrate ditetrafluoroborate
PubChem CID 177615
SMILES O.O.O.O.O.O.[Ni++].F[B-](F)(F)F.F[B-](F)(F)F

Specifications

Physical Form Lump
Quantity 100 g
UN Number UN3077
Solubility Information Soluble in water.
IUPAC Name nickel(2+) bis(tetrafluoroboranuide) hexahydrate
Formula Weight 340.40 (232.30 Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Density 1.5 g/mL
Chemical Name or Material Nickel (II) tetrafluoroborate hexahydrate
Hazard Category H314-H350
Hazard Statement GHS H Statement
H334-H350-H360-H372-H341-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P264b-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 238-753-4
RTECSNumber QR6650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.