Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel Iron Molybdenum foil, 0.102mm (0.004in) thick, annealed

Catalog No. p-7059662
Change view
Click to view available options
Quantity:
150 x 300 mm
300 x 300 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA450924K 150 x 300 mm
AA45092HB 300 x 300 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA450924K Supplier Thermo Scientific Chemicals Supplier No. 0450924K
Only null left
Add to Cart

Form: approx. 40.1g/150 x 300mm | Ni:Fe:Mo

It is most commonly used as a high temperature, corrosion resistant metal alloy in steel, cast iron and super alloys for the military and defense industry, semiconductor industry and specialty machine shops.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nickel Iron Molybdenum foil
Name Note 0.102mm (0.004 in.) thick, Annealed
Avg. Mol. Wt. or Mol. Wt. Range Ni:Fe:Mo; 80:15:4.2 wt%
Quantity 150 x 300 mm
Weight ≈40.1g/150x300mm
Form Foil
Molecular Formula Ni:Fe:Mo
MDL Number MFCD09910537
Solubility Information Insoluble in water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.