Learn More
Glass Expansion Nickel Jet Sampler Cone for Thermo Scientific Element 1/Element 2/Neptune/Neoma ICP-MS (1260630)
SDP
Catalog No. 501907709
Encompass_Preferred
Nickel Jet Sampler Cone (1260630) ¿ Suitable for Thermo/Finnigan Neoma MC-ICP-MS. Ideal for high sensitivity applications with low matrix containing samples. Suitable for most common applications (e.g., routine aqueous samples, <5% acid matrix, non-HF, non-organic). Not recommended for samples containing hydrofluoric acid (HF). Use a new Graphite Sampler Cone Gasket (501907717) each time the cone is re-installed, even if it¿s the same cone. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For lowest boron background and semiconductor applications, we recommend a platinum cone, see catalog number 501907711.For standard Nickel Sampler with Copper Core, see catalog number 501907705.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7709 Supplier Glass Expansion Supplier No. TF1005NI
Nickel Jet Sampler Cone (1260630) ¿ Suitable for Thermo/Finnigan Neoma MC-ICP-MS. Ideal for high sensitivity applications with low matrix containing samples. Suitable for most common applications (e.g., routine aqueous samples, <5% acid matrix, non-HF, non-organic). Not recommended for samples containing hydrofluoric acid (HF). Use a new Graphite Sampler Cone Gasket (501907717) each time the cone is re-installed, even if it¿s the same cone. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For lowest boron background and semiconductor applications, we recommend a platinum cone, see catalog number 501907711.For standard Nickel Sampler with Copper Core, see catalog number 501907705.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001