Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel Nitrate Hexahydrate (Crystalline/Certified), Fisher Chemical

Catalog No. N62500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
N62-500 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. N62-500

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 13478-00-7
Molecular Formula H12N2NiO12
Molecular Weight (g/mol) 290.79
MDL Number MFCD00149805
InChI Key AOPCKOPZYFFEDA-UHFFFAOYSA-N
Synonym nickel nitrate hexahydrate, nickel ii nitrate hexahydrate, nickel dinitrate hexahydrate, nickel 2+ dinitrate hexahydrate, unii-xbt61wlt1j, nickelous nitrate hexahydrate, nickel 2+ nitrate, hexahydrate, xbt61wlt1j, nitric acid, nickel 2+ salt, hexahydrate, nickel ii nitrate, hexahydrate 1:2:6
PubChem CID 61630
IUPAC Name nickel(2+) hexahydrate dinitrate
SMILES O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Boiling Point 137°C
Melting Point 56.7°C
Color Blue-Green
pH 5
Physical Form Solid
Quantity 500 g
Linear Formula Ni(NO3)2·6H2O
Formula Weight 290.81
Grade Certified
Identification Pass Test
Packaging Glass Bottle
Chemical Name or Material Nickel Nitrate Hexahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.