Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel nitrate, Matrix Modifier Solution, Specpure™

Catalog No. AA39043AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA39043AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA39043AE Supplier Thermo Scientific Chemicals Supplier No. 039043AE
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7697-37-2 | HNO3 | 63.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7697-37-2 , 13478-00-7 , 7732-18-5
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Color Green
Physical Form Liquid
Quantity 100 mL
Concentration or Composition (by Analyte or Components) Nitric acid: 2.0%; Nickel(II) nitrate hexahydrate: 1.0%; Water: 97.0%
Appearance Green liquid
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Chemical Name or Material Nickel nitrate
Hazard Category H290-H315-H317-H319-H334-H341-H350i-H360D-H373
Hazard Statement Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
Precautionary Statement P201-P202-P234-P260-P264b-P272-P280-P281-P285-P302+P352-P304+P341-P305+P351+P338-P308+P313-P333+P313-P342+P311-P362-P390-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.